siRNA Details
| Gene Symbol: Oxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130163.1 | NP_001123635.1 | 14 | 2892 | ||
| NM_001130164.1 | NP_001123636.1 | 6 | 1245 | |||
| NM_001130165.1 | NP_001123637.1 | 14 | 2987 | |||
| NM_001130166.1 | NP_001123638.1 | 17 | 3332 | |||
| NM_130885.2 | NP_570955.1 | 15 | 2973 | |||
| GenBank mRNA | AF324899.1 | - | 15 | 2972 | ||
| AF333985.1 | - | 14 | 2892 | |||
| AK158703.1 | - | 17 | 3314 | |||
| BC003927.1 | - | 8 | 1482 | |||
| BC089183.1 | - | 14 | 2982 | 5321255 | ||
| BC098491.1 | - | 16 | 3234 | 30086857 | ||

Oxr1
Genomic Map