siRNA Details
| Gene Symbol: Card14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130886.3 | NP_570956.1 | 4, 5 | 670 | ||
| GenBank mRNA | AF363457.1 | - | 4, 5 | 649 | ||
| AK086176.1 | - | 5, 6 | 919 | |||
| BC004692.1 | - | 1, 2 | 105 | |||
| BC029102.1 | - | 4, 5 | 670 | 3979883 | ||

Card14
Genomic Map