siRNA Details
| Gene Symbol: Nxf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130888.1 | NP_570958.1 | 8 | 689 | ||
| GenBank mRNA | AB178901.1 | - | 9 | 828 | ||
| AJ305317.1 | - | 5 | 358 | |||
| AJ305318.1 | - | 5 | 358 | |||
| AY260550.1 | - | 8 | 689 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nxf7| Gene Symbol: Nxf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130888.1 | NP_570958.1 | 8 | 689 | ||
| GenBank mRNA | AB178901.1 | - | 9 | 828 | ||
| AJ305317.1 | - | 5 | 358 | |||
| AJ305318.1 | - | 5 | 358 | |||
| AY260550.1 | - | 8 | 689 | |||