siRNA Details
| Gene Symbol: Nxf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130888.1 | NP_570958.1 | 11 | 1016 | ||
| GenBank mRNA | AB178901.1 | - | 12 | 1155 | ||
| AJ305317.1 | - | 7 | 577 | |||
| AJ305318.1 | - | 8 | 685 | |||
| AY260550.1 | - | 11 | 1016 | |||

Nxf7
Genomic Map