siRNA Details
| Gene Symbol: Znrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168621.1 | NP_001162092.1 | 5 | 1975 | ||
| NM_001168622.1 | NP_001162093.1 | 3 | 2990 | |||
| NM_001168623.1 | NP_001162094.1 | 5 | 1174 | |||
| NM_133206.3 | NP_573469.1 | 4 | 3086 | |||
| GenBank mRNA | AF378525.1 | - | 5 | 1680 | ||
| AK030575.1 | - | 3 | 2137 | |||
| AK078819.1 | - | 5 | 1200 | |||
| BC086770.1 | - | 4 | 2885 | 5720382 | ||

Znrf1
Genomic Map