siRNA Details
| Gene Symbol: TRDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006073.3 | NP_006064.2 | 23, 24 | 1790 | ||
| GenBank mRNA | CR749591.1 | - | 12, 13 | 498 | ||
| U18985.1 | - | 23, 24 | 1494 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRDN| Gene Symbol: TRDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006073.3 | NP_006064.2 | 23, 24 | 1790 | ||
| GenBank mRNA | CR749591.1 | - | 12, 13 | 498 | ||
| U18985.1 | - | 23, 24 | 1494 | |||