siRNA Details
| Gene Symbol: Acbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133225.3 | NP_573488.2 | 8 | 1605 | ||
| GenBank mRNA | AF022770.3 | - | 8 | 1606 | ||
| AK030371.1 | - | 8 | 1606 | |||
| AK168799.1 | - | 8 | 1579 | |||
| BC060602.1 | - | 8 | 1560 | 5706182 | ||

Acbd3
Genomic Map