siRNA Details
| Gene Symbol: Hook2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167991.1 | NP_001161463.1 | 22 | 2336 | ||
| NM_133255.2 | NP_573518.2 | 22 | 2339 | |||
| GenBank mRNA | AK030599.1 | - | 24 | 2355 | ||
| AK042048.1 | - | 22 | 2327 | |||
| AK136069.1 | - | 23 | 2261 | |||
| AK145768.1 | - | 22 | 2307 | |||
| AK166954.1 | - | 22 | 2316 | |||
| AK169073.1 | - | 22 | 2307 | |||
| BC002226.1 | - | 14 | 1548 | |||
| BC019486.1 | - | 22 | 2315 | 4211990 | ||
| BC026609.1 | - | 14 | 1499 | |||
| BC029967.1 | - | 22 | 2280 | |||
| BC055881.1 | - | 22 | 2282 | 3590336 | ||
| BC080744.1 | - | 22 | 2329 | 30471369 | ||

Hook2
Genomic Map