siRNA Details
| Gene Symbol: Oosp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133353.3 | NP_579931.1 | 2 | 196 | ||
| GenBank mRNA | AB050982.2 | - | 2 | 189 | ||
| AB086437.1 | - | 2 | 179 | |||
| AF420487.1 | - | 2 | 201 | |||
| AK033038.1 | - | 2 | 199 | |||
| AK162132.1 | - | 2 | 199 | |||
| BC100313.1 | - | 2 | 175 | 30936071 | ||

Oosp1
Genomic Map