siRNA Details
| Gene Symbol: Rab31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133685.2 | NP_598446.2 | 5 | 534 | ||
| GenBank mRNA | AB232617.1 | - | 5 | 326 | ||
| AK051270.1 | - | 5 | 454 | |||
| AK156677.1 | - | 5 | 535 | |||
| AK159095.1 | - | 5 | 423 | |||
| BC013063.1 | - | 5 | 463 | 3497938 | ||

Rab31
Genomic Map