siRNA Details
| Gene Symbol: Cops8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133805.3 | NP_598566.3 | 2, 3 | 277 | ||
| GenBank mRNA | AF482000.1 | - | 2, 3 | 738 | ||
| AK079111.1 | - | 2, 3 | 211 | |||
| AK079522.1 | - | 2, 3 | 225 | |||
| AK088323.1 | - | 2, 3 | 197 | |||
| AK090013.1 | - | 2, 3 | 206 | |||
| AK145899.1 | - | 2, 3 | 204 | |||
| AK162502.1 | - | 2, 3 | 212 | |||
| AK195692.1 | - | 1, 2 | 52 | |||
| BC017690.1 | - | 2, 3 | 245 | 4238947 | ||
| BC021488.1 | - | 2, 3 | 236 | 5344864 | ||
| BC024421.1 | - | 2, 3 | 230 | 4987584 | ||

Cops8
Genomic Map