siRNA Details
| Gene Symbol: Cops8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133805.3 | NP_598566.3 | 5 | 503 | ||
| GenBank mRNA | AF482000.1 | - | 5 | 964 | ||
| AK079111.1 | - | 5 | 437 | |||
| AK079522.1 | - | 5 | 451 | |||
| AK088323.1 | - | 5 | 423 | |||
| AK090013.1 | - | 5 | 432 | |||
| AK142989.1 | - | 4 | 1472 | |||
| AK145899.1 | - | 5 | 430 | |||
| AK162502.1 | - | 5 | 438 | |||
| BC017690.1 | - | 5 | 471 | 4238947 | ||
| BC021488.1 | - | 5 | 462 | 5344864 | ||
| BC024421.1 | - | 5 | 456 | 4987584 | ||

Cops8
Genomic Map