siRNA Details
| Gene Symbol: Cops8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133805.3 | NP_598566.3 | 8 | 1526 | ||
| GenBank mRNA | AF482000.1 | - | 8 | 1987 | ||
| AK079111.1 | - | 8 | 1460 | |||
| AK079522.1 | - | 8 | 1474 | |||
| AK088323.1 | - | 8 | 1446 | |||
| AK090013.1 | - | 8 | 1455 | |||
| AK142989.1 | - | 7 | 2495 | |||
| AK145899.1 | - | 8 | 1453 | |||
| AK162502.1 | - | 8 | 1461 | |||
| AK185916.1 | - | 1 | 298 | |||
| AK207412.1 | - | 1 | 293 | |||
| AK212344.1 | - | 1 | 291 | |||
| AK214410.1 | - | 1 | 290 | |||
| AK215258.1 | - | 1 | 293 | |||
| AK218596.1 | - | 1 | 291 | |||
| BC017690.1 | - | 8 | 1493 | 4238947 | ||
| BC021488.1 | - | 8 | 1484 | 5344864 | ||
| BC024421.1 | - | 8 | 1478 | 4987584 | ||

Cops8
Genomic Map