siRNA Details
| Gene Symbol: Mmadhc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133839.2 | NP_598600.1 | 8 | 834 | ||
| GenBank mRNA | BC002253.1 | - | 8 | 834 | 3493526 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmadhc| Gene Symbol: Mmadhc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133839.2 | NP_598600.1 | 8 | 834 | ||
| GenBank mRNA | BC002253.1 | - | 8 | 834 | 3493526 | |