siRNA Details
| Gene Symbol: C8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133882.2 | NP_598643.1 | 7 | 1156 | ||
| GenBank mRNA | AB077304.1 | - | 7 | 1078 | ||
| AK050313.1 | - | 7 | 1157 | |||
| BC022129.1 | - | 5 | 787 | |||
| BC096382.1 | - | 7 | 1130 | 5102825 | ||

C8b
Genomic Map