siRNA Details
Gene Symbol: Sptssa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134054.2 | NP_598815.2 | 2 | 799 | ||
GenBank mRNA | AK003171.1 | - | 2 | 736 | ||
AK003754.1 | - | 1 | 195 | |||
AK008904.1 | - | 2 | 756 | |||
AK018380.1 | - | 2 | 776 | |||
AK137543.1 | - | 2 | 774 | |||
BC022674.1 | - | 2 | 725 | 4236922 |
