siRNA Details
| Gene Symbol: Rnf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146025.1 | NP_001139497.1 | 9 | 1439 | ||
| NM_001146026.1 | NP_001139498.1 | 9 | 1364 | |||
| NM_001146027.1 | NP_001139499.1 | 9 | 1230 | |||
| NM_134064.2 | NP_598825.2 | 9 | 1372 | |||
| GenBank mRNA | AK147349.1 | - | 9 | 1365 | ||
| AK160090.1 | - | 9 | 1223 | |||
| AK160115.1 | - | 9 | 1223 | |||
| AK161651.1 | - | 9 | 1354 | |||
| AK171943.1 | - | 9 | 1221 | |||
| AK172257.1 | - | 9 | 1222 | |||
| BC017630.1 | - | 9 | 1165 | 4507289 | ||
| BC035548.1 | - | 9 | 1187 | 4221284 | ||

Rnf44
Genomic Map