siRNA Details
| Gene Symbol: Rnf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146025.1 | NP_001139497.1 | 11 | 2318 | ||
| NM_001146026.1 | NP_001139498.1 | 11 | 2243 | |||
| NM_001146027.1 | NP_001139499.1 | 11 | 2109 | |||
| NM_134064.2 | NP_598825.2 | 11 | 2251 | |||
| GenBank mRNA | AK147349.1 | - | 11 | 2244 | ||
| AK160090.1 | - | 11 | 2106 | |||
| AK160115.1 | - | 11 | 2106 | |||
| AK161651.1 | - | 11 | 2235 | |||
| AK171943.1 | - | 11 | 2104 | |||
| AK172257.1 | - | 11 | 2105 | |||
| BC017630.1 | - | 11 | 2044 | 4507289 | ||
| BC035548.1 | - | 11 | 2066 | 4221284 | ||

Rnf44
Genomic Map