siRNA Details
| Gene Symbol: Epdr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134065.4 | NP_598826.3 | 3 | 1509 | ||
| GenBank mRNA | AK034083.1 | - | 3 | 1478 | ||
| AK038733.1 | - | 3 | 1484 | |||
| AK046185.1 | - | 3 | 1484 | |||
| AK160413.1 | - | 3 | 1444 | |||
| AK166252.1 | - | 3 | 1451 | |||
| BC014708.1 | - | 3 | 1464 | 4223134 | ||

Epdr1
Genomic Map