siRNA Details
| Gene Symbol: AW209491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104646.1 | NP_001098116.1 | 2 | 1831 | ||
| NM_134067.4 | NP_598828.3 | 2 | 1898 | |||
| GenBank mRNA | AK030506.1 | - | 1880 | |||
| AK050507.1 | - | 2 | 1880 | |||
| BC016098.1 | - | 1 | 1792 | 4910571 | ||
| BU530510.1 | - | 1 | 240 | |||

AW209491
Genomic Map