siRNA Details
| Gene Symbol: AW209491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030506.1 | - | 2070 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AW209491| Gene Symbol: AW209491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030506.1 | - | 2070 | |||