siRNA Details
| Gene Symbol: Kctd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111028.1 | NP_001104498.1 | 10, 11 | 1165 | ||
| NM_001285933.1 | NP_001272862.1 | 10, 11 | 1292 | |||
| NM_134073.2 | NP_598834.2 | 9, 10 | 1015 | |||
| GenBank mRNA | AK087312.1 | - | 10, 11 | 1039 | ||
| AK158922.1 | - | 9, 10 | 1017 | |||
| AK166501.1 | - | 9, 10 | 851 | |||
| BC011302.1 | - | 10, 11 | 1106 | |||
| CK618608.1 | - | 3, 4 | 310 | |||

Kctd9
Genomic Map