siRNA Details
| Gene Symbol: Chmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134078.4 | NP_598839.2 | 4 | 779 | ||
| GenBank mRNA | AK211372.1 | - | 2 | 125 | ||
| BC006648.1 | - | 1 | 216 | |||
| BC024115.1 | - | 4 | 767 | 5100379 | ||
| BC033365.1 | - | 4 | 750 | 5367653 | ||

Chmp7
Genomic Map