siRNA Details
| Gene Symbol: Chmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134078.4 | NP_598839.2 | 10 | 2568 | ||
| GenBank mRNA | AK011732.1 | - | 1 | 949 | ||
| AK031882.1 | - | 1 | 869 | |||
| AK179878.1 | - | 1 | 209 | |||
| AK182423.1 | - | 1 | 210 | |||
| AK190385.1 | - | 1 | 210 | |||
| AK191247.1 | - | 1 | 210 | |||
| AK196156.1 | - | 1 | 210 | |||
| BC006648.1 | - | 4 | 3231 | |||
| BC024115.1 | - | 10 | 2556 | 5100379 | ||
| BC033365.1 | - | 8 | 2998 | 5367653 | ||

Chmp7
Genomic Map