siRNA Details
Gene Symbol: Slc38a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166456.1 | NP_001159928.1 | 2 | 390 | ||
NM_001166458.1 | NP_001159930.1 | 2 | 597 | |||
NM_134086.4 | NP_598847.2 | 2 | 1083 | |||
GenBank mRNA | AF184240.2 | - | 1 | 175 | ||
AK029189.1 | - | 2 | 260 | |||
AK034130.1 | - | 2 | 1085 | |||
AK035098.1 | - | 1 | 102 | |||
AK047459.1 | - | 2 | 614 | |||
AK049173.1 | - | 2 | 297 | |||
AK049294.1 | - | 2 | 363 | |||
AK050914.1 | - | 2 | 298 | |||
AK081724.1 | - | 2 | 599 | |||
AK082683.1 | - | 2 | 298 | |||
AK162612.1 | - | 2 | 448 | |||
AK165188.1 | - | 2 | 650 | |||
AK199869.1 | - | 1 | 14 | |||
BC030378.1 | - | 2 | 299 | 5375319 | ||
BC066815.1 | - | 2 | 260 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
