siRNA Details
| Gene Symbol: MAT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005911.5 | NP_005902.1 | 3, 4 | 610 | ||
| GenBank mRNA | AK291126.1 | - | 3, 4 | 418 | ||
| AK293841.1 | - | 3, 4 | 420 | |||
| AK297758.1 | - | 3, 4 | 313 | |||
| AK308591.1 | - | 3, 4 | 313 | |||
| AK308758.1 | - | 3, 4 | 278 | |||
| AK309338.1 | - | 3, 4 | 418 | |||
| AK316411.1 | - | 3, 4 | 568 | |||
| BC001686.2 | - | 3, 4 | 407 | 3010820 | ||
| BC001854.2 | - | 3, 4 | 407 | 3010820 | ||
| BI549760.1 | - | 3, 4 | 429 | |||
| DC322518.1 | - | 3, 4 | 451 | |||
| DQ083239.1 | - | 3, 4 | 346 | |||
| X68836.1 | - | 3, 4 | 365 | |||

MAT2A
Genomic Map