siRNA Details
| Gene Symbol: Fbxo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134099.2 | NP_598860.2 | 2 | 552 | ||
| GenBank mRNA | BC036385.1 | - | 2 | 517 | ||
| BC040086.1 | - | 2 | 552 | 3495299 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo4| Gene Symbol: Fbxo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134099.2 | NP_598860.2 | 2 | 552 | ||
| GenBank mRNA | BC036385.1 | - | 2 | 517 | ||
| BC040086.1 | - | 2 | 552 | 3495299 | ||