siRNA Details
| Gene Symbol: Trip10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242389.1 | NP_001229318.1 | 15 | 2099 | ||
| NM_001242390.1 | NP_001229319.1 | 15 | 2096 | |||
| NM_001242391.1 | NP_001229320.1 | 14 | 1928 | |||
| NM_134125.4 | NP_598886.1 | 14 | 1931 | |||
| GenBank mRNA | AK088909.1 | - | 14 | 1878 | ||
| AK149902.1 | - | 14 | 1900 | |||
| AY081142.1 | - | 15 | 2047 | |||
| BC003249.1 | - | 14 | 1933 | 3500118 | ||
| CD348583.1 | - | 4 | 674 | |||
| CJ230382.1 | - | 2 | 360 | |||

Trip10
Genomic Map