siRNA Details
| Gene Symbol: Fbxo38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134136.3 | NP_598897.2 | 2 | 271 | ||
| GenBank mRNA | AK031096.1 | - | 2 | 221 | ||
| AK031347.1 | - | 2 | 202 | |||
| AK089046.1 | - | 2 | 243 | |||
| AY267463.1 | - | 2 | 204 | |||
| BC056348.1 | - | 2 | 223 | 5721407 | ||
| BY131284.1 | - | 2 | 272 | |||

Fbxo38
Genomic Map