siRNA Details
| Gene Symbol: Cyp2c50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC011222.1 | - | 6 | 837 | 4195223 | |
| BC051050.1 | - | 5 | 668 | 5052536 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2c50| Gene Symbol: Cyp2c50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC011222.1 | - | 6 | 837 | 4195223 | |
| BC051050.1 | - | 5 | 668 | 5052536 | ||