siRNA Details
| Gene Symbol: AI837181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256515.1 | NP_001243444.1 | 2 | 957 | ||
| NM_134149.2 | NP_598910.2 | 2 | 943 | |||
| GenBank mRNA | AK077296.1 | - | 1 | 679 | ||
| BC017542.1 | - | 2 | 824 | 4526963 | ||
| BC024516.1 | - | 2 | 944 | 4984074 | ||
| U64690.1 | - | 2 | 822 | |||

AI837181
Genomic Map