siRNA Details
| Gene Symbol: Lpxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134152.3 | NP_598913.1 | 2 | 153 | ||
| GenBank mRNA | AB053936.2 | - | 2 | 205 | ||
| AB071194.1 | - | 2 | 178 | |||
| AK039922.1 | - | 2 | 183 | |||
| AK077085.1 | - | 2 | 189 | |||
| BC026563.1 | - | 2 | 119 | 5065405 | ||

Lpxn
Genomic Map