siRNA Details
| Gene Symbol: Vmn1r237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134200.1 | NP_598961.1 | 1 | 653 | ||
| GenBank mRNA | BC125543.1 | - | 1 | 707 | 40130058 | |
| BC138066.1 | - | 1 | 707 | 8861085 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r237| Gene Symbol: Vmn1r237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134200.1 | NP_598961.1 | 1 | 653 | ||
| GenBank mRNA | BC125543.1 | - | 1 | 707 | 40130058 | |
| BC138066.1 | - | 1 | 707 | 8861085 | ||