siRNA Details
Gene Symbol: Vmn1r233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134202.1 | NP_598963.1 | 1 | 733 | ||
GenBank mRNA | BC115528.1 | - | 1 | 729 | 40101376 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134202.1 | NP_598963.1 | 1 | 733 | ||
GenBank mRNA | BC115528.1 | - | 1 | 729 | 40101376 |