siRNA Details
Gene Symbol: Vmn1r198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134220.1 | NP_598981.1 | 1 | 131 | ||
GenBank mRNA | BC130236.1 | - | 1 | 135 | 40142042 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vmn1r198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134220.1 | NP_598981.1 | 1 | 131 | ||
GenBank mRNA | BC130236.1 | - | 1 | 135 | 40142042 |