siRNA Details
| Gene Symbol: Timd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161355.1 | NP_001154827.1 | 9 | 1766 | ||
| NM_001161356.1 | NP_001154828.1 | 9 | 1539 | |||
| NM_134249.5 | NP_599010.4 | 9 | 1648 | |||
| GenBank mRNA | AK145630.1 | - | 9 | 1528 | ||
| AK163225.1 | - | 9 | 1527 | |||
| AK166877.1 | - | 9 | 1520 | |||
| BC028829.1 | - | 9 | 1611 | 5102129 | ||

Timd2
Genomic Map