siRNA Details
| Gene Symbol: Havcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134250.2 | NP_599011.2 | 7 | 1976 | ||
| GenBank mRNA | AF450241.1 | - | 7 | 1964 | ||
| AK170775.1 | - | 8 | 1799 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Havcr2| Gene Symbol: Havcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134250.2 | NP_599011.2 | 7 | 1976 | ||
| GenBank mRNA | AF450241.1 | - | 7 | 1964 | ||
| AK170775.1 | - | 8 | 1799 | |||