siRNA Details
| Gene Symbol: Slc26a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134420.4 | NP_599252.2 | 2558 | |||
| GenBank mRNA | AB099881.1 | - | 1719 | |||
| AF248494.1 | - | 1904 | ||||
| AK029238.1 | - | 3480 | ||||
| AK046993.1 | - | 3818 | ||||
| AK053088.1 | - | 2209 | ||||
| AK163670.1 | - | 2559 | ||||
| AK163671.1 | - | 2559 | ||||
| AY032863.1 | - | 1863 | ||||
| AY049076.1 | - | 1818 | ||||
| BC028856.1 | - | 1882 | 4165725 | |||
| CN692543.1 | - | 378 | ||||

Slc26a6
Genomic Map