siRNA Details
Gene Symbol: AVPR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000707.3 | NP_000698.1 | 2 | 1474 | ||
GenBank mRNA | AF030512.1 | - | 2 | 1439 | ||
AF101726.1 | - | 2 | 1439 | |||
D31833.1 | - | 2 | 1523 | |||
L37112.1 | - | 2 | 1409 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
