siRNA Details
| Gene Symbol: Aldh7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127338.1 | NP_001120810.1 | 17 | 1605 | ||
| NM_138600.4 | NP_613066.2 | 15 | 1411 | |||
| GenBank mRNA | AK004991.1 | - | 17 | 1539 | ||
| AK145873.1 | - | 15 | 1354 | |||
| AK160117.1 | - | 15 | 1338 | |||
| AK169195.1 | - | 15 | 1354 | |||
| BC012407.1 | - | 15 | 1373 | 4191228 | ||

Aldh7a1
Genomic Map