siRNA Details
| Gene Symbol: Fam50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138607.3 | NP_613073.2 | 8 | 848 | ||
| GenBank mRNA | AK008402.1 | - | 8 | 849 | ||
| BC026128.1 | - | 7 | 1221 | |||
| BC125297.1 | - | 7 | 714 | 40129812 | ||
| Y18505.1 | - | 8 | 813 | |||

Fam50a
Genomic Map