siRNA Details
| Gene Symbol: Hps4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138646.3 | NP_619587.3 | 4 | 781 | ||
| GenBank mRNA | AK081489.1 | - | 4 | 741 | ||
| AK129418.1 | - | 3 | 668 | |||
| AK147954.1 | - | 3 | 1224 | |||
| AK171871.1 | - | 4 | 750 | |||
| AK187573.1 | - | 1 | 14 | |||
| AY043415.1 | - | 3 | 301 | |||
| BC063055.1 | - | 3 | 473 | 5701206 | ||

Hps4
Genomic Map