siRNA Details
| Gene Symbol: Nlgn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163387.1 | NP_001156859.1 | 5 | 1516 | ||
| NM_138666.3 | NP_619607.2 | 7 | 2543 | |||
| GenBank mRNA | AK122433.1 | - | 7 | 2544 | ||
| AK138209.1 | - | 2 | 377 | |||
| BC098461.1 | - | 5 | 1516 | 30099449 | ||

Nlgn1
Genomic Map