siRNA Details
| Gene Symbol: Luc7l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138680.2 | NP_619621.2 | 7, 8 | 1191 | ||
| GenBank mRNA | AF318301.1 | - | 7, 8 | 1116 | ||
| AK078853.1 | - | 35 | ||||
| BC002314.1 | - | 2, 3 | 126 | |||
| BC020058.1 | - | 8, 9 | 916 | 4009362 | ||
| BC040793.1 | - | 6, 7 | 683 | |||
| BC054449.1 | - | 6, 7 | 1134 | |||
| BC056354.1 | - | 7, 8 | 1189 | 6811460 | ||
| BC056970.1 | - | 7, 8 | 1189 | 6811460 | ||

Luc7l2
Genomic Map