siRNA Details
| Gene Symbol: Hlcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139145.4 | NP_631884.1 | 10 | 3625 | ||
| GenBank mRNA | AB066227.1 | - | 9 | 3485 | ||
| AK052424.1 | - | 11 | 3921 | |||
| AK158212.1 | - | 11 | 3738 | |||
| AK168925.1 | - | 7 | 2522 | |||
| BC050090.1 | - | 10 | 3626 | 30012868 | ||

Hlcs
Genomic Map