siRNA Details
| Gene Symbol: Tmem63a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144794.2 | NP_659043.1 | 23 | 2952 | ||
| GenBank mRNA | AK049360.1 | - | 23 | 2949 | ||
| AK144899.1 | - | 23 | 3005 | |||
| AK170559.1 | - | 23 | 2968 | |||
| AK173027.1 | - | 23 | 2945 | |||
| BC014795.1 | - | 23 | 2925 | 3961992 | ||
| BC019442.1 | - | 23 | 2952 | 4036849 | ||

Tmem63a
Genomic Map