siRNA Details
| Gene Symbol: TUBA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270399.1 | NP_001257328.1 | 4 | 2302 | ||
| NM_001270400.1 | NP_001257329.1 | 4 | 1830 | |||
| NM_006009.3 | NP_006000.2 | 4 | 1742 | |||
| GenBank mRNA | AF141347.1 | - | 4 | 1476 | ||
| AF141348.1 | - | 1 | 1029 | |||
| AK057842.1 | - | 4 | 1496 | |||
| AK289483.1 | - | 4 | 1669 | |||
| AK297775.1 | - | 3 | 689 | |||
| BC006468.1 | - | 4 | 1451 | 3528520 | ||
| BC050637.2 | - | 4 | 1481 | 6050536 | ||
| K00557.1 | - | 2 | 1070 | |||

TUBA1A
Genomic Map