siRNA Details
| Gene Symbol: Rnf185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290472.1 | NP_001277401.1 | 7 | 2460 | ||
| NM_001290473.1 | NP_001277402.1 | 6 | 2441 | |||
| NM_145355.5 | NP_663330.2 | 8 | 2579 | |||
| GenBank mRNA | AK034108.1 | - | 8 | 1949 | ||
| AK052436.1 | - | 7 | 1869 | |||
| AK053358.1 | - | 7 | 1826 | |||
| AK077638.1 | - | 7 | 1791 | |||
| AK149640.1 | - | 7 | 1812 | |||
| AK165895.1 | - | 1882 | ||||
| AK167455.1 | - | 7 | 1777 | |||
| AK168904.1 | - | 7 | 1814 | |||

Rnf185
Genomic Map