siRNA Details
| Gene Symbol: Zbtb7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145356.3 | NP_663331.1 | 4 | 1082 | ||
| GenBank mRNA | AK045522.1 | - | 4 | 1046 | ||
| AK156372.1 | - | 2 | 750 | |||
| BC018187.1 | - | 2 | 779 | 4208891 | ||
| BC070424.1 | - | 4 | 1082 | 6837447 | ||

Zbtb7c
Genomic Map