siRNA Details
| Gene Symbol: Wfdc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145369.3 | NP_663344.2 | 4 | 775 | ||
| GenBank mRNA | AK075896.1 | - | 3 | 465 | ||
| AK131905.1 | - | 4 | 723 | |||
| AK131906.1 | - | 4 | 708 | |||
| AY669505.1 | - | 4 | 774 | |||
| BC019734.1 | - | 4 | 775 | 3987617 | ||

Wfdc5
Genomic Map